Orders LinkContactUsLinkComponent
Lab Consumables
Lab Consumables
17, Non-calibrated loop, Ø 2 mm, 26 gal., with aluminium insulated handle, 100×1, bulk
Katalogové číslo 612-2674
Dodavatel:   Copan
Lab Consumables
Katalogové číslo 612-2674
Dodavatel:  Copan
Lab Consumables
Lab Consumables
17, Non-calibrated loop, Ø 2 mm, 26 gal., with aluminium insulated handle, 100×1, bulk
Přejít na:
Price
3 980,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
A variety of high-quality lab consumables to meet laboratories everyday needs. We offer a wide range of transfer pipets, loops, spreaders, tubes, caps, and droppers meeting the highest quality standards. Since we produce them with our in-house technology, we can easily customise them upon customer’s request.
Streaking:
Loops and spreaders
Needles, spreaders, and inoculating loops fitting a multitude of different tasks
- Lubricants, oils, and electrostatic charges free
- Certified calibration
- Hexagonal loop shaft for easy handling
- Evans blue dye accuracy certification
Specifikace
  • Balené:
    100×1, bulk
  • Popis:
    Non-calibrated loop, Ø 2 mm, 26 gal., with aluminium insulated handle
  • Sekvence:
    17