Orders LinkContactUsLinkComponent
Lab Consumables
Lab Consumables
16, Non-calibrated loop, Ø 5 mm, 26 gal., 2500×1, bulk
Catalog # 612-2671
Supplier:   Copan
Lab Consumables
Catalog # 612-2671
Supplier:  Copan
Lab Consumables
Lab Consumables
16, Non-calibrated loop, Ø 5 mm, 26 gal., 2500×1, bulk
Price
4 510,00 Kč
Log in for availability
1 * 25 KS
Product Details & Documents
A variety of high-quality lab consumables to meet laboratories everyday needs. We offer a wide range of transfer pipets, loops, spreaders, tubes, caps, and droppers meeting the highest quality standards. Since we produce them with our in-house technology, we can easily customise them upon customer’s request.
Streaking:
Loops and spreaders
Needles, spreaders, and inoculating loops fitting a multitude of different tasks
- Lubricants, oils, and electrostatic charges free
- Certified calibration
- Hexagonal loop shaft for easy handling
- Evans blue dye accuracy certification
Specifications
  • Balené:
    2500×1, bulk
  • Popis:
    Non-calibrated loop, Ø 5 mm, 26 gal.
  • Sekvence:
    16