Orders LinkContactUsLinkComponent
Trypsinising flasks
Trypsinising flasks
04, 2000 ml
Katalogové číslo WITG5.507.306
Dodavatel:   witeg Labortechnik
Trypsinising flasks
Katalogové číslo WITG5.507.306
Dodavatel:  witeg Labortechnik
Trypsinising flasks
Trypsinising flasks
04, 2000 ml
Přejít na:
Price
2 610,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Trypsinising flasks, graduated, designed for converting tissue fragments and homogenates into cell suspensions by trypsin digestion of connective proteins. Flutes in flask enhance vigorous agitation.
Specifikace
  • Kapacita:
    2000 ml
  • Sekvence:
    04