Orders LinkContactUsLinkComponent
Adhesive plasters, Salvequick
Adhesive plasters, Salvequick
22, Salvequick XL fabric plasters refill, incl. 15 sterile plasters (87×58 mm)
Katalogové číslo 118-0028
Dodavatel:   ORKLA CARE
Adhesive plasters, Salvequick
Katalogové číslo 118-0028
Dodavatel:  ORKLA CARE
Adhesive plasters, Salvequick
Adhesive plasters, Salvequick
22, Salvequick XL fabric plasters refill, incl. 15 sterile plasters (87×58 mm)
Přejít na:
Price
2 030,00 Kč
Přihlásit se pro dostupnost
1 * 6 KS
Product Details & Documents
These plaster refills are available in different versions: Made from plastic or fabric, blue detectable and for sensitive skin.
  • In combination with dispenser: Complete, easy to use system
  • Plasters are always available, ready to use
  • Plasters are allergy-tested
  • Each adhesive bandage is individually wrapped and sterile
Detectable plasters are specially designed for use in the food industry. The plasters are detectable by most metal detectors used within the food industry. The blue colour also makes the plasters easy to see.
Sensitive plasters are white and made of polyurethane. They are water repellent and suitable for sensitive skin.
Documents
Specifikace
  • Popis:
    Salvequick XL fabric plasters refill, incl. 15 sterile plasters (87×58 mm)
  • Sekvence:
    22