[EN]POROLYTE SOLUTION 1 * 50 mL
Katalogové číslo METR6.2318.000
Dodavatel: METROHM
Frequently Bought Together
Standard vodivosti 10 µS/cm (25°C; 0,000027 mol/l; KCl)
Dodavatel: Mettler - Toledo
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Dodavatel: VWR Chemicals
pH buffer solutions, certified (DKD)
Dodavatel: Mettler - Toledo
pH buffer solutions
Dodavatel: Mettler - Toledo
FREEZING POINT STD CRYOSCOPE 621 100ML 1 * 1 KS
Dodavatel: REAGECON
N-NITROSO-ETHYL-ISOPROPYLAMINE 100 µG/ML 1 * 1 mL
Dodavatel: EHRENSTORFER
Sulfamethoxazole, Reference Standard
Dodavatel: USP
Reagents for chloride determination using Lovibond® MD photometers
Dodavatel: Lovibond
PAH-MIX 14 10 NG/µL 1 * 10 mL
Dodavatel: EHRENSTORFER
Iron, plasma standard solution, Fe 1000 µg/ml, Specpure®
Dodavatel: Thermo Fisher Scientific