Orders LinkContactUsLinkComponent
Flat flange lids with 5 necks
NS 29/32 (Centre), 3×29/32 angled and 1×14/23 straight NS
  201-1143
Flat flange lids with 5 necks
  201-1143
Flat flange lids with 5 necks
Flat flange lids with 5 necks
NS 29/32 (Centre), 3×29/32 angled and 1×14/23 straight NS
12 890,00 Kč
1 * 1 KS
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat laboratory flange (DN). Straight middle neck, three angled side necks and one straight side neck with standard ground joints.
  • Hrdlo:
    NS 29/32 (Centre)
  • Boční hrdla:
    3×29/32 angled and 1×14/23 straight NS