[EN]SQW.1ML INSERT.CAP.SI/PTFE.351.9MM. 1 * 1 KS
Katalogové číslo LAPH08200911
Dodavatel: LA PHA PACK
[EN]SQW.1ML INSERT.CAP.SI/PTFE.351.9MM. 1 * 1 KS
Katalogové číslo LAPH08200911
Dodavatel: LA PHA PACK
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR® Advanced, Návleky na boty, protiskluzové
Dodavatel: VWR Collection
VWR®, Short Thread Vials ND9
Dodavatel: VWR Collection
VWR®, Screw Neck Vials ND8
Dodavatel: VWR Collection
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Pláště laboratorní, KIMTECH SCIENCE* A7 P+
Dodavatel: KIMBERLY CLARK
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Screw Neck Vials ND24
Dodavatel: VWR Collection
VWR®, Snap Cap Vials
Dodavatel: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals