Conductivity cells, ScienceLine
32, 4-pole conductivity cell, sensor 4 platinum rings, temp. sensor NTC 30 kOhm, LF 713 T, 1 µS/cm - 2000 mS/cm, 1,0 cm-1, −30...+135 °C, Sklo, 12×120 mm, Fixed 1 m cable with 8 pole plug
Katalogové číslo 663-0282
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
These high-end conductivity cells are ideal for the monitoring and control of plants as well as the examination of ultrapure water in quality control labs.
- Models with SMEK plug head can be connected to diverse instrument types
- Glass platinum electrodes LF713T (4-pole) and LF913T (5-pole) allow the determination of conductivity in organic media
Specifikace
- Vodivost:1 µS/cm - 2000 mS/cm
- Připojení:Fixed 1 m cable with 8 pole plug
- Ø×D:12×120 mm
- Type:LF 713 T
- Popis:4-pole conductivity cell, sensor 4 platinum rings, temp. sensor NTC 30 kOhm
- Materiál:Sklo
- Sekvence:32
- Tepl.:−30...+135 °C
- Konstanta cely:1,0 cm-1