Orders LinkContactUsLinkComponent
Drying tubes, bend
Drying tubes, bend
19/26 NS
Katalogové číslo WITG1.660.019
Dodavatel:   witeg Labortechnik
Drying tubes, bend
Katalogové číslo WITG1.660.019
Dodavatel:  witeg Labortechnik
Drying tubes, bend
Drying tubes, bend
19/26 NS
Přejít na:
Price
672,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3. Angled at 75° with ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.
Specifikace
  • Rozměry zábrusu:
    19/26 NS