Orders LinkContactUsLinkComponent
Drying tubes, bend
Drying tubes, bend
24/29 NS
Catalog # WITG1.660.024
Supplier:   witeg Labortechnik
Drying tubes, bend
Catalog # WITG1.660.024
Supplier:  witeg Labortechnik
Drying tubes, bend
Drying tubes, bend
24/29 NS
Price
763,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3. Angled at 75° with ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.
Specifications
  • Rozměry zábrusu:
    24/29 NS