Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
03, Electrode cable, for connecting electrodes with Metrohm plug-in head G to Metrohm instruments (socket F), 3 m
Catalog # METR6.2104.040
Supplier:   METROHM
Electrode Cables
Catalog # METR6.2104.040
Supplier:  METROHM
Electrode Cables
03, Electrode cable, for connecting electrodes with Metrohm plug-in head G to Metrohm instruments (socket F), 3 m
Price
7 100,00 Kč
Log in for availability
1 * 3 m
Product Details & Documents
Specifications
  • Popis:
    Electrode cable, for connecting electrodes with Metrohm plug-in head G to Metrohm instruments (socket F)
  • Délka:
    3 m
  • Sekvence:
    03