Orders LinkContactUsLinkComponent
Gravity hydrometers
Gravity hydrometers
002, ASTM hydrometer, 29…41 °API, 330 mm
Katalogové číslo AMARH825006
Dodavatel:   Amarell
Gravity hydrometers
Katalogové číslo AMARH825006
Dodavatel:  Amarell
Gravity hydrometers
Gravity hydrometers
002, ASTM hydrometer, 29…41 °API, 330 mm
Přejít na:
Price
1 620,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These gravity hydrometers are designed with - and without - thermometer.
  • Paper scale
  • Type designation
  • High precision
NOTE: Until 10th October 2017 it is still allowed to use mercury thermometers, if they are exclusively intended to be used for tests as per standards, which require the usage of mercury thermometers.
Certifications: ASTM
Specifikace
  • Popis:
    ASTM hydrometer
  • Délka:
    330 mm
  • Sekvence:
    002
  • Rozsah:
    29…41 °API