Orders LinkContactUsLinkComponent
Watchmakers' magnifiers
Watchmakers' magnifiers
004, Magnifier with dual lens, 15×, 13 mm
Katalogové číslo ESCH1130
Dodavatel:   ESCHENBACH OPTIK
Watchmakers' magnifiers
Katalogové číslo ESCH1130
Dodavatel:  ESCHENBACH OPTIK
Watchmakers' magnifiers
Watchmakers' magnifiers
004, Magnifier with dual lens, 15×, 13 mm
Přejít na:
Price
1 200,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite
Specifikace
  • Ø:
    13 mm
  • Zvětšení:
    15×
  • Popis:
    Magnifier with dual lens
  • Sekvence:
    004