Orders LinkContactUsLinkComponent
Glass beads
Glass beads
05, Glass beads, solid, 5 mm
Catalog # 201-0279
Glass beads
Catalog # 201-0279
Glass beads
Glass beads
05, Glass beads, solid, 5 mm
Price
660,00 Kč
Log in for availability
1 * 1 kg
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
  • Ø:
    5 mm
  • Popis:
    Glass beads, solid
  • Sekvence:
    05