Orders LinkContactUsLinkComponent
Glass beads
Glass beads
03, 3 mm
Catalog # 201-0538
Glass beads
Catalog # 201-0538
Glass beads
Glass beads
03, 3 mm
Price
630,00 Kč
Log in for availability
1 * 1 kg
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
  • Ø:
    3 mm
  • Sekvence:
    03