Orders LinkContactUsLinkComponent
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
2000 ml, 29/32 NS, 19/26 NS
Katalogové číslo WITG0.770.002
Dodavatel:   witeg Labortechnik
Flasks, round bottom, with three parallel necks
Katalogové číslo WITG0.770.002
Dodavatel:  witeg Labortechnik
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
2000 ml, 29/32 NS, 19/26 NS
Přejít na:
Price
3 510,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Specifikace
  • Kapacita:
    2000 ml
  • Boční hrdla:
    19/26 NS
  • Centrální hrdlo:
    29/32 NS