Orders LinkContactUsLinkComponent
Comparator kits for water testing
Molybdate Test Kit, Model MO-2, 0 - 0,8/0 - 40 mg/l, 100, 1 KIT
  HACH1419301
 :   Hach
Comparator kits for water testing
  HACH1419301
 :  Hach
 :  
Comparator kits for water testing
Comparator kits for water testing
Molybdate Test Kit, Model MO-2, 0 - 0,8/0 - 40 mg/l, 100, 1 KIT
 :  
13 990,00 Kč
1 * 1 KIT
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
  • Bal.:
    1 KIT
  • Popis:
    Molybdate Test Kit, Model MO-2
  • Testů v kitu:
    100
  • Rozsah:
    0 - 0,8/0 - 40 mg/l
Accessories
  •  
    Popis
    Bal.
  • HACH11032H
    Ammonium molybdate reagent for phosphate determinations, 100 ml
    1
    1 * 1 KS - 865,00 Kč