KIT LUMIGLO A+B (6X120ML) 1 * 6 KS
Catalog # KPLI54-61-01
Supplier: SERACARE LIFE SCIENCES
Frequently Bought Together
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR®, Pipety Pasteurovy, Standard Line
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
E.Z.N.A.® Plasmid Mini kit I
Supplier: OMEGA BIO-TEK
VWR®, Roztěrky, tvar L, jednorázové
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Nádoby na vzorky se šroubovacím uzávěrem
Supplier: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF