Orders LinkContactUsLinkComponent
Rectangular cover glasses, Menzel Gläser
21, Cover glasses, 24×36 mm, #1,5
Katalogové číslo 630-2871
Dodavatel:   Epredia
Rectangular cover glasses, Menzel Gläser
Katalogové číslo 630-2871
Dodavatel:  Epredia
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
21, Cover glasses, 24×36 mm, #1,5
Přejít na:
Price
2 350,00 Kč
Přihlásit se pro dostupnost
1 * 1.000 KS
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles or flaws
  • Made of pre-washed pure white Hydrolytic Class 1 glass
Specifikace
  • Počet:
    #1,5
  • Popis:
    Cover glasses
  • D׊:
    24×36 mm
  • Sekvence:
    21