Orders LinkContactUsLinkComponent
Syringes, SampleLock™
Syringes, SampleLock™
03, 22, 5000 µl, 51 mm, 2, 1005 RN
Catalog # HAMI81556
Supplier:   HAMILTON BONADUZ
Syringes, SampleLock™
Catalog # HAMI81556
Supplier:  HAMILTON BONADUZ
Syringes, SampleLock™
Syringes, SampleLock™
03, 22, 5000 µl, 51 mm, 2, 1005 RN
Price
9 950,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
  • Not autoclavable, gas sterilisable
  • An easy-to-use twist valve
  • A positive rear stop prevents loss of sample and plunger blowout
  • Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
Specifications
  • Rozchod:
    22
  • Type:
    1005 RN
  • Typ špičky:
    2
  • Objem:
    5000 µl
  • Délka:
    51 mm
  • Sekvence:
    03
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • HAMI35081
    02
    Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock termination
    Log in for availability
    1 * 1 KS - 1 460,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • HAMI35080
    01
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Tip (LT) termination
    Log in for availability
    1 * 1 KS - 1 200,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • HAMI35083
    03
    Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Lock (TLL) termination
    Log in for availability
    1 * 1 KS - 1 300,00 Kč
Product Information
  • Catalog #
    Sekvence
    Rozchod
    Délka
    Availability
    Price
  • HAMI7784-02
    37
    22
    51 mm
    Log in for availability
    1 * 6 KS - 5 920,00 Kč
Product Information
  • Catalog #
    Sekvence
    Rozchod
    Délka
    Availability
    Price
  • HAMI7779-01
    28
    22
    51 mm
    Log in for availability
    1 * 6 KS - 2 050,00 Kč
Product Information
  • Catalog #
    Sekvence
    Rozchod
    Délka
    Availability
    Price
  • HAMI7784-01
    36
    22s
    51 mm
    Log in for availability
    1 * 6 KS - 6 030,00 Kč
Product Information
  • Catalog #
    Sekvence
    Rozchod
    Délka
    Availability
    Price
  • HAMI7779-03
    27
    22s
    51 mm
    Log in for availability
    1 * 6 KS - 2 050,00 Kč
Product Information
  • Catalog #
    Sekvence
    Rozchod
    Délka
    Availability
    Price
  • HAMI7780-03
    31
    22s
    51 mm
    Log in for availability
    1 * 6 KS - 1 620,00 Kč
Product Information
  • Catalog #
    Sekvence
    Rozchod
    Délka
    Availability
    Price
  • HAMI7780-04
    32
    22
    51 mm
    Log in for availability
    1 * 6 KS - 1 570,00 Kč