Syringes, SampleLock™
Product Details & Documents
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
- Not autoclavable, gas sterilisable
- An easy-to-use twist valve
- A positive rear stop prevents loss of sample and plunger blowout
- Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
Specifikace
- Rozchod:22
- Type:1025 RN
- Typ špičky:2
- Objem:25000 µl
- Délka:51 mm
- Sekvence:05
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Removable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock terminationRemovable needle adapter, to convert a large hub removable needle termination into a Female Luer Lock terminationHAMI3508102Přihlásit se pro dostupnost1 * 1 KS - 1 460,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Tip (LT) terminationRemovable needle adapter, to convert a large hub removable needle termination into a Male Luer Tip (LT) terminationHAMI3508001Přihlásit se pro dostupnost1 * 1 KS - 1 200,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Removable needle adapter, to convert a large hub removable needle termination into a Male Luer Lock (TLL) terminationRemovable needle adapter, to convert a large hub removable needle termination into a Male Luer Lock (TLL) terminationHAMI3508303Přihlásit se pro dostupnost1 * 1 KS - 1 300,00 Kč
Informace o produktu
- Katalogové čísloSekvenceRozchodDélkaDostupnostPrice
Informace o produktu
- Katalogové čísloSekvenceRozchodDélkaDostupnostPrice
Informace o produktu
- Katalogové čísloSekvenceRozchodDélkaDostupnostPrice
Informace o produktu
- Katalogové čísloSekvenceRozchodDélkaDostupnostPrice
Informace o produktu
- Katalogové čísloSekvenceRozchodDélkaDostupnostPrice
Informace o produktu
- Katalogové čísloSekvenceRozchodDélkaDostupnostPrice