ContactUsLinkComponent
NUT, PEEK, FINGERTIGHT, 1/16'', 10-32 1 * 5 KS
Catalog # JOURJR-5570-5
Supplier:  VICI JOUR RESEARCH
NUT, PEEK, FINGERTIGHT, 1/16'', 10-32 1 * 5 KS
Catalog # JOURJR-5570-5
Supplier:  VICI JOUR RESEARCH
Frequently Bought Together