Orders LinkContactUsLinkComponent
Slide boxes, SlideFile™ Junior
Slide boxes, SlideFile™ Junior
Modrá, PS, 200, 82×140×86 mm
Catalog # 631-1905
Supplier:   Simport Scientific
Slide boxes, SlideFile™ Junior
Catalog # 631-1905
Supplier:  Simport Scientific
Slide boxes, SlideFile™ Junior
Slide boxes, SlideFile™ Junior
Modrá, PS, 200, 82×140×86 mm
Price
2 780,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Boxes are impact-resistant and stackable for space efficient storage.
  • Barcodes can be read through the clear lid
  • Number of slides can be doubled by storing two in a slot
  • Tinted hinged cover makes the content easy to see
 : Include a slide box and a removable tray.
Specifications
  • Počet sklíček:
    200
  • Barva:
    Modrá
  • Materiál:
    PS
  • Š×H×V:
    82×140×86 mm