Orders LinkContactUsLinkComponent
Vacuum stopcocks
14, Vacuum stopcocks, one way, right-angled, 8,0 mm, 24 NS
Katalogové číslo 189-2124
Dodavatel:   Lenz Laborglas
Vacuum stopcocks
Katalogové číslo 189-2124
Dodavatel:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
14, Vacuum stopcocks, one way, right-angled, 8,0 mm, 24 NS
Přejít na:
Price
2 580,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Specifikace
  • Ø otvoru:
    8,0 mm
  • Popis:
    Vacuum stopcocks, one way, right-angled
  • Sekvence:
    14
  • Kohout:
    24 NS