Orders LinkContactUsLinkComponent
Face shields, Bionic™
Anti-scratch, anti-fog, PC
  111-0134
Face shields, Bionic™
  111-0134
Face shields, Bionic™
Face shields, Bionic™
Anti-scratch, anti-fog, PC
1 540,00 Kč
1 * 1 KS
This innovative face shield provides protection from extreme mechanical hazards. It can be worn in combination with safety spectacles or respiratory protection.
  • With extended protection for brow and chin
  • Locking mechanism for added safety
  • Extra soft padding with washable or replaceable sweatband
  • Various adjustments for best comfort
Weight: 355 g.
EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Certifications: EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
  • Type:
    Anti-scratch, anti-fog
  • Materiál:
    PC
Accessories
  •  
    Popis
    Materiál
  • 111-0740
    Replacement visor, uncoated
    PC
    1 * 1 KS - 263,00 Kč
  •  
    Popis
    Materiál
  • 111-2160
    Replacement visor, uncoated
    Acetate
    1 * 1 KS - 429,00 Kč
  •  
    Popis
    Materiál
  • 111-1293
    Replacement visor, anti-scratch/anti-fog
    PC
    1 * 1 KS - 679,00 Kč
  •  
    Popis
  • 113-8053
    Replacement sweatband
    1 * 1 KS - 154,00 Kč