Orders LinkContactUsLinkComponent
Stopcocks, one-way
16, 4,0 mm, 14,5 NS
Catalog # LENZ02161404
Supplier:   Lenz Laborglas
Stopcocks, one-way
Catalog # LENZ02161404
Supplier:  Lenz Laborglas
Stopcocks, one-way
Stopcocks, one-way
16, 4,0 mm, 14,5 NS
Price
1 070,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
 : DIN 12554.
Specifications
  • Ø otvoru:
    4,0 mm
  • Sekvence:
    16
  • Kohout:
    14,5 NS