[EN]VIAL KIT,4ML,SCREW NECK,CAP,PTFE SEP 1 * 100 KS
Katalogové číslo LAPH13281434
Dodavatel: LA PHA PACK
[EN]VIAL KIT,4ML,SCREW NECK,CAP,PTFE SEP 1 * 100 KS
Katalogové číslo LAPH13281434
Dodavatel: LA PHA PACK
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Dodavatel: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Dodavatel: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
VWR®, Snap Cap Vials
Dodavatel: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR®, Stojany na zkumavky, skládací a stohovatelné
Dodavatel: VWR Collection
VWR®, Short Thread Vials ND9
Dodavatel: VWR Collection
Voda USP, JP, Ph. Eur., purified
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Screw caps for certified screw top vials
Dodavatel: Agilent