Orders LinkContactUsLinkComponent
Test kits, COD cell test, Spectroquant®, Supelco®
24, Kyvetový test COD*⁷⁻¹, 15−300 mg/L, Chromosulfurická oxidace, Prove, Nova 30/60, Multy, Move, 25 Tests
  1.14895.0001
 :   Merck
Test kits, COD cell test, Spectroquant®, Supelco®
  1.14895.0001
 :  Merck
 :  
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
24, Kyvetový test COD*⁷⁻¹, 15−300 mg/L, Chromosulfurická oxidace, Prove, Nova 30/60, Multy, Move, 25 Tests
 :  
4 630,00 Kč
1 * 25 Tests
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and wastewater
*8 COD cell tests fully comply with ISO 15705.
  • Chemická metoda:
    Chromosulfurická oxidace
  • Bal.:
    25 Tests
  • Popis:
    Kyvetový test COD*⁷⁻¹
  • Sekvence:
    24
  • Pro jednotku:
    Prove, Nova 30/60, Multy, Move
  • Rozsah:
    15−300 mg/L
  • Cat. No.:
    1.14895.0001
  • M-Clarity quality level:
    MQ100
Accessories
  •  
    Sekvence
    Popis
    Bal.
  • 1.15955.0001
    02
    Nástavce dávkovací k CHSK analýze mořské vody 1.17059.0001
    1
    1 * 1 KS - 6 790,00 Kč
  •  
    Sekvence
    Typ balení
    Bal.
  • 1.06733.0501
    3645
    Plastová láhev
    500 g
    1 * 500 g - 2 780,00 Kč