Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
Separating funnels, pear shaped
500 ml, 24/29 NS
Katalogové číslo 511-0756
Dodavatel:   SciLabware
Separating funnels, pear shaped
Katalogové číslo 511-0756
Dodavatel:  SciLabware
Separating funnels, pear shaped
Separating funnels, pear shaped
500 ml, 24/29 NS
Přejít na:
Price
3 180,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.
Specifikace
  • Velikost objímky:
    24/29 NS
  • Kapacita:
    500 ml