Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
Separating funnels, pear shaped
100 ml, 19/26 NS
Katalogové číslo 511-0754
Dodavatel:   SciLabware
Separating funnels, pear shaped
Katalogové číslo 511-0754
Dodavatel:  SciLabware
Separating funnels, pear shaped
Separating funnels, pear shaped
100 ml, 19/26 NS
Přejít na:
Price
2 940,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.
Specifikace
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    100 ml