Orders LinkContactUsLinkComponent
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
03, 22×32 mm, #1
Catalog # 631-0133
Supplier:   VWR Collection
VWR® Rectangular Cover Glasses
Catalog # 631-0133
Supplier:  VWR Collection
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
03, 22×32 mm, #1
Price
1 360,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: Sklíčka jsou balená v plastových krabičkách opatřených víčkem s panty.
Specifications
  • Počet:
    #1
  • D׊:
    22×32 mm
  • Sekvence:
    03