Orders LinkContactUsLinkComponent
Mops, PowerHead™
Mops, PowerHead™
08, PowerHead™ wide pore foam, 356 mm
Katalogové číslo 129-0525
Dodavatel:   MICRONOVA
Mops, PowerHead™
Katalogové číslo 129-0525
Dodavatel:  MICRONOVA
Mops, PowerHead™
Mops, PowerHead™
08, PowerHead™ wide pore foam, 356 mm
Přejít na:
Price
539,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Mop head is constructed of polyester-covered foam mop heads are available both nonsterile and sterile for critical cleaning. Absorbent foam mop head is for pharmaceutical production and buffer areas. Wide-pore foam mop head and dense urethane foam mop head are for cleanroom support and maintenance areas.
  • Stainless steel PowerHead™ handles hold and wring the various mop heads
  • Autoclavable
  • A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles
Informace pro objednávání: All components are ordered separately.
Specifikace
  • Popis:
    PowerHead™ wide pore foam, 356 mm
  • Sekvence:
    08