Orders LinkContactUsLinkComponent
Mops, PolyGen™
Mops, PolyGen™
Mop head, sterile
Katalogové číslo 129-0520
Dodavatel:   MICRONOVA
Mops, PolyGen™
Katalogové číslo 129-0520
Dodavatel:  MICRONOVA
Mops, PolyGen™
Mops, PolyGen™
Mop head, sterile
Price
2 370,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The dual-action abrasive mesh is specially adapted to pharmaceutical and biotech projects. Heads used directly with the fibreglass/PP handle or attached to the universal handle via the quick disconnect adapter.
  • Mop can clean to a dry finish or apply disinfectants requiring extended drying time
  • Special herringbone weave ensures no raw edges to guarantee low particulation
  • Low-linting polyester looped strands
  • Reusable, launderable, and autoclavable
Ordering information: Mop heads and handle must be ordered separately.
Specifikace
  • Popis:
    Mop head, sterile
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    04
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 430,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Délka
    Dostupnost
    Price
  • Prodlužovací nerezová tyč
    Prodlužovací nerezová tyč
    129-0551
    01
    965 - 1829 mm
    Přihlásit se pro dostupnost
    1 * 1 KS - 7 840,00 Kč