Orders LinkContactUsLinkComponent
Mops, PolyGen™
Mops, PolyGen™
Mop head
Catalog # 129-0167
   Waste Reducing
Supplier:   MICRONOVA
Mops, PolyGen™
Catalog # 129-0167
Supplier:  MICRONOVA
   Waste Reducing
Mops, PolyGen™
Mops, PolyGen™
Mop head
Price
2 110,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
The dual-action abrasive mesh is specially adapted to pharmaceutical and biotech projects. Heads used directly with the fibreglass/PP handle or attached to the universal handle via the quick disconnect adapter.
  • Mop can clean to a dry finish or apply disinfectants requiring extended drying time
  • Special herringbone weave ensures no raw edges to guarantee low particulation
  • Low-linting polyester looped strands
  • Reusable, launderable, and autoclavable
Ordering information: Mop heads and handle must be ordered separately.
Specifications
  • Popis:
    Mop head
  • Cat. No.:
    129-0167
  • Environmentally Preferable:
    Waste Reducing
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 129-0170
    04
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    Log in for availability
    1 * 1 KS - 3 430,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Délka
    Availability
    Price
  • 129-0551
    01
    Prodlužovací nerezová tyč
    965 - 1829 mm
    Log in for availability
    1 * 1 KS - 7 840,00 Kč