Orders LinkContactUsLinkComponent
Mops, MicroMop™
Mops, MicroMop™
02, Mop head, NovaLite™
Katalogové číslo 129-0506
   Waste Reducing
Dodavatel:   MICRONOVA
Mops, MicroMop™
Katalogové číslo 129-0506
Dodavatel:  MICRONOVA
   Waste Reducing
Mops, MicroMop™
Mops, MicroMop™
02, Mop head, NovaLite™
Price
4 320,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
MicroMop™ are ideal for quick and efficient cleaning of large surface areas. Suitable for wafer processing, electronics, pharmaceutical, and biotech applications. All mop heads attach to stainless steel universal handle via the orange polypropylene quick disconnect adapter. For less critical applications, choose the fibreglass/polypropylene handle with attached adapter.
  • Low-linting, quick drying PVA is ideal for high-tech applications. Can be laundered
  • Polyester has good wicking capability, can be autoclaved and used with strong disinfectants
  • Lite™ is sturdy latex/rayon blend for controlled environments and cleanroom support areas; mop head can be autoclaved for medical and sterile applications
Informace pro objednávání: All components must be ordered separately.
Specifikace
  • Popis:
    Mop head, NovaLite™
  • Sekvence:
    02
  • Cat. No.:
    129-0506
  • Environmentally Preferable:
    Waste Reducing
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    04
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 430,00 Kč