Orders LinkContactUsLinkComponent
Accessories for mops micromop™ LoopMop™ and PolyGen™
Accessories for mops micromop™ LoopMop™ and PolyGen™
04, MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
Katalogové číslo 129-0170
Dodavatel:   MICRONOVA
Accessories for mops micromop™ LoopMop™ and PolyGen™
Katalogové číslo 129-0170
Dodavatel:  MICRONOVA
Accessories for mops micromop™ LoopMop™ and PolyGen™
Accessories for mops micromop™ LoopMop™ and PolyGen™
04, MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
Přejít na:
Price
3 430,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Universal electropolished stainless steel handles with a quick disconnect connection that works with the entire range of Micronova adapters, tools and mop heads for simplified and versatile cleaning.
  • Universal handles work with all Quick Disconnect tools and mop adapters
  • 304 electropolished stainless steel with polyketone components
  • Strong, hard-wearing connections for long lasting use
  • Available in a range of fixed or extendable lengths from 8 in (20 cm) to 10 ft (3 m)
  • Quick Disconnect end attaches directly to QD mops, tools and adapters
  • SSA-3 adapter connects QD handles to plastic threaded mops, tools and adapters
  • Use the Adjustable Angle Adapters to reach over, under and around obstacles in the cleanroom
  • Good compatibility with disinfectants
  • Autoclavable
The Micronova Stainless Steel Universal Handles attach directly to the stainless steel Quick Disconnect (QD series) adapters and tools. Use the SSA-3 Quick Disconnect Adapter to attach to the plastic ACME threaded (QP series) adapters and tools. The Adjustable Angle Adapters can be used to create an adjustable angled shape between a handle and mop adapter to clean above and below fixtures, as well as, inside isolators and containment hoods.
Specifikace
  • Popis:
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
  • Sekvence:
    04