Orders LinkContactUsLinkComponent
Pipeclay triangles
Pipeclay triangles
02, Pipeclay stems on stout wire, 63 mm
Catalog # MURRL026/03
Supplier:   MURRAY S
Pipeclay triangles
Catalog # MURRL026/03
Supplier:  MURRAY S
Pipeclay triangles
02, Pipeclay stems on stout wire, 63 mm
Price
901,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Pipeclay stems on stout wire.
Specifications
  • Popis:
    Pipeclay stems on stout wire, 63 mm
  • Sekvence:
    02