Orders LinkContactUsLinkComponent
Watchmaker forceps
Watchmaker forceps
Watchmaker No. 7, curved, 115 mm
Katalogové číslo 231-0435
Dodavatel:   MURRAY S
Watchmaker forceps
Katalogové číslo 231-0435
Dodavatel:  MURRAY S
Watchmaker forceps
Watchmaker forceps
Watchmaker No. 7, curved, 115 mm
Přejít na:
Price
301,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stainless steel. Ideal for small-scale dissection applications or precise part placement.
  • Very fine points
  • Smooth flat handle
  • Available straight or curved
Specifikace
  • Type:
    Watchmaker No. 7, curved
  • Délka:
    115 mm