Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Safety spectacles, Silium

Safety spectacles, Silium

02, Anti-UV (5-2,5), anti-fog, anti-scratch, Stříbrná, Smoke PC
Katalogové číslo 111-1646
Dodavatel:   Bollé Safety
Safety spectacles, Silium
Katalogové číslo 111-1646
Dodavatel:  Bollé Safety
Safety spectacles, Silium
Safety spectacles, Silium

02, Anti-UV (5-2,5), anti-fog, anti-scratch, Stříbrná, Smoke PC

Přejít na:
Price
428,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents

These slim and attractively designed spectacles feature a metal, nickel-free frame and a curved lens.

  • Flexible temples bend up to 160%, equipped with comfort pads
  • Flexible fittings in the nose bridge offer extra comfort

Weight: 34 g

EN 166, EN 170

 : EN 166, EN 170

Documents

Specifikace
  • Type:
    Anti-UV (5-2,5), anti-fog, anti-scratch
  • Barva:
    Stříbrná
  • Sekvence:
    02
  • Čočky/objektiv:
    Smoke PC
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.