Orders LinkContactUsLinkComponent
Maintenance sorbents
Maintenance sorbents
Chemicals sorbent roll, 117 l/roll, 46 m×400 mm
Katalogové číslo MMMAMB2001
Dodavatel:   3M
Maintenance sorbents
Katalogové číslo MMMAMB2001
Dodavatel:  3M
Maintenance sorbents
Maintenance sorbents
Chemicals sorbent roll, 117 l/roll, 46 m×400 mm
Přejít na:
Price
3 990,00 Kč
Přihlásit se pro dostupnost
1 * 1 Roll
Product Details & Documents
For use in production facilities and maintenance shops to handle routine leaks, drips, spray, and fluid splashes from nonaggressive liquids. Nonaggressive liquids are those used in daily operations which do not require the use of personal protective equipment. These include oils, coolants, cutting fluids, aqueous-based liquids, and petroleum-based products.
Unlike clay granules, these sorbents are not messy and can be used on equipment. This allows placement at the source of the leak, preventing spread to floor. The various sizes and styles available also permit use under, around, or for wiping down equipment, as well as in areas where foot and vehicle traffic is an issue. At full capacity, they take on darker grey colour, making the user aware that they are ready to be changed. Sorbents are made from synthetic fibers and incorporate the 3M™ patented microbundle construction.
Upozornení: Nevhodné pro nebezpečné kapaliny. Všechny produkty 3M™ musí být používány v souladu s předpisy OSHA a pokyny pro uživatele, upozornění a omezení jsou součástí každého produktu.
Specifikace
  • Popis:
    Chemicals sorbent roll
  • D׊:
    46 m×400 mm
  • Savost:
    117 l/roll