Orders LinkContactUsLinkComponent
Dressing scissors, straight
Dressing scissors, straight
Probe/sharp, 125 mm
Catalog # 233-0016
Supplier:   MURRAY S
Dressing scissors, straight
Catalog # 233-0016
Supplier:  MURRAY S
Dressing scissors, straight
Dressing scissors, straight
Probe/sharp, 125 mm
Price
289,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel. These straight scissors can be used for a variety of applications in medical areas, laboratories and microscopy.
Specifications
  • Type:
    Probe/sharp
  • Délka:
    125 mm