Joint clips for conical joints, POM, Quickfit®
08, Okrová, 40/38
Katalogové číslo 201-3032
Dodavatel: SciLabware
Přejít na:
Product Details & Documents
POM, coloured. Polyacetal clips for retaining glassware having Quickfit® conical joints.
- Easy to fit and remove
- Can be used at temperatures up to +80 °C
- Clips are colour coded for easy identification
Specifikace
- Pro NZ:40/38
- Barva:Okrová
- Sekvence:08