Orders LinkContactUsLinkComponent
Accessories for screw thread joints
Accessories for screw thread joints
6,0 - 7,0 mm, 28
Katalogové číslo 201-0695
Dodavatel:   SciLabware
Accessories for screw thread joints
Katalogové číslo 201-0695
Dodavatel:  SciLabware
Accessories for screw thread joints
Accessories for screw thread joints
6,0 - 7,0 mm, 28
Přejít na:
Price
2 560,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifikace
  • Ø otvoru:
    6,0 - 7,0 mm
  • Závit:
    28