Orders LinkContactUsLinkComponent
Accessories for screw thread joints
Accessories for screw thread joints
10,5 - 11,5 mm, 24
Catalog # 201-0690
Supplier:   SciLabware
Accessories for screw thread joints
Catalog # 201-0690
Supplier:  SciLabware
Accessories for screw thread joints
Accessories for screw thread joints
10,5 - 11,5 mm, 24
Price
1 190,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifications
  • Ø otvoru:
    10,5 - 11,5 mm
  • Závit:
    24