Accessories for screw thread joints
7,0 - 8,5 mm, 18
Katalogové číslo 201-0689
Dodavatel: SciLabware
Přejít na:
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifikace
- Ø otvoru:7,0 - 8,5 mm
- Závit:18
For use with Quickfit® glass screw thread.