Orders LinkContactUsLinkComponent
Accessories for screw thread joints
Accessories for screw thread joints
10,5 - 11,5 mm, 28
Katalogové číslo BIBBQC28/13
Dodavatel:   SciLabware
Accessories for screw thread joints
Katalogové číslo BIBBQC28/13
Dodavatel:  SciLabware
Accessories for screw thread joints
Accessories for screw thread joints
10,5 - 11,5 mm, 28
Přejít na:
Price
2 270,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifikace
  • Ø otvoru:
    10,5 - 11,5 mm
  • Závit:
    28