Orders LinkContactUsLinkComponent
Accessories for screw thread joints
Accessories for screw thread joints
7,0 - 8,5 mm, 18
Katalogové číslo BIBBQC18/11
Dodavatel:   SciLabware
Accessories for screw thread joints
Katalogové číslo BIBBQC18/11
Dodavatel:  SciLabware
Accessories for screw thread joints
Accessories for screw thread joints
7,0 - 8,5 mm, 18
Přejít na:
Price
1 770,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifikace
  • Ø otvoru:
    7,0 - 8,5 mm
  • Závit:
    18