Orders LinkContactUsLinkComponent
Accessories for screw thread joints
Accessories for screw thread joints
6,0 -7,0 mm, 13
Catalog # 201-0895
Supplier:   SciLabware
Accessories for screw thread joints
Catalog # 201-0895
Supplier:  SciLabware
Accessories for screw thread joints
Accessories for screw thread joints
6,0 -7,0 mm, 13
Price
1 190,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifications
  • Ø otvoru:
    6,0 -7,0 mm
  • Závit:
    13