Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, blunt, 115 mm
Catalog # 232-0030
Supplier:   MURRAY S
Forceps
Catalog # 232-0030
Supplier:  MURRAY S
Forceps
Forceps
Straight, blunt, 115 mm
Price
174,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
  • Blunt, narrow flutes
  • For dissecting and general applications
Specifications
  • Type:
    Straight, blunt
  • Délka:
    115 mm