Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Unisette™ M506
Biopsy processing/embedding cassettes, Unisette™ M506
09, Purpurová
Katalogové číslo 720-1564
Dodavatel:   Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M506
Katalogové číslo 720-1564
Dodavatel:  Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M506
Biopsy processing/embedding cassettes, Unisette™ M506
09, Purpurová
Přejít na:
Price
5 470,00 Kč
Přihlásit se pro dostupnost
1 * 1.500 KS
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Integral lid
  • Front face inclined at an angle of 45° so that the cassette can be used with certain types of cassette labelling devices
  • Available in 11 non cytotoxic, non metallic colours and three fluorescent colours
Packaging: Each case contains three boxes of 500 cassettes each.
Specifikace
  • Barva:
    Purpurová
  • Sekvence:
    09